Product Details

SNP ID
rs62466836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93424507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGAATGAAAAGATGAATAATA[C/T]ATTTCTTTAGAAAAATATGCAAATA
Phenotype
MIM: 114131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 3557 UTR 3 NP_001158209.1
NM_001164738.1 3557 UTR 3 NP_001158210.1
NM_001742.3 3557 UTR 3 NP_001733.1

View Full Product Details