Product Details

SNP ID
rs61756141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:104126891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTTATTATGTCAAAGTTCACC[A/G]TCCTAAAAACAAAAACAGATAATAT
Phenotype
MIM: 602331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ORC5 PubMed Links

Gene Details

Gene
ORC5
Gene Name
origin recognition complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002553.3 1408 Missense Mutation ACG,ATG T422M NP_002544.1
NM_181747.3 1408 Intron NP_859531.1
XM_011516273.2 1408 Intron XP_011514575.1

View Full Product Details