Product Details
- SNP ID
-
rs55988854
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:108564972 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGTAATTCTGGATGTTGATGG[A/G]GCACATCTGTGTTAACTATATTTTT
- Phenotype
-
MIM: 602634
MIM: 612534
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111825882] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB9
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B9
There are no transcripts associated with this gene.
- Gene
- THAP5
- Gene Name
- THAP domain containing 5
View Full Product Details