Product Details
- SNP ID
-
hCV88888097
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130270770 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGTGGTAACTTCAATTTTGGGGC[C/T]TACCATACCCTGGAAGAGGTAGGTG
- Phenotype
-
MIM: 600688
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPA2
PubMed Links
Gene Details
- Gene
- CPA2
- Gene Name
- carboxypeptidase A2
There are no transcripts associated with this gene.
View Full Product Details