Product Details

SNP ID
rs55989609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135670009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGTTGTGTACTGCTCTGTGCC[A/G]GTCTAGACTTCTCTTTTGGGGCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf73 PubMed Links

Gene Details

Gene
C7orf73
Gene Name
chromosome 7 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130929.1 Intron NP_001124401.1

View Full Product Details