Product Details

SNP ID
rs61456957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39002658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTTTTAACTCACTTCCCTGTA[A/C]TACAATTACTATTATGAAAAGTGTG
Phenotype
MIM: 602713 MIM: 610081
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAM9 PubMed Links

Gene Details

Gene
ADAM9
Gene Name
ADAM metallopeptidase domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003816.2 Intron NP_003807.1
XM_011544682.2 Intron XP_011542984.1
XM_017013942.1 Intron XP_016869431.1
Gene
TM2D2
Gene Name
TM2 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details