Product Details

SNP ID
rs56156301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103135576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCTGTGGGAATATGCCTTTTGG[A/G]ATCTATTCTCACCTCTTATTCAGTC
Phenotype
MIM: 606602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAALC PubMed Links

Gene Details

Gene
BAALC
Gene Name
brain and acute leukemia, cytoplasmic
There are no transcripts associated with this gene.

Gene
BAALC-AS2
Gene Name
BAALC antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details