Product Details

SNP ID
rs62522491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:141398997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCAGTCCCCTTCCCACGGGAC[C/T]GGTGCTCCCGGCTGCTCTCCTGGCC
Phenotype
MIM: 606449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTP4A3 PubMed Links

Gene Details

Gene
PTP4A3
Gene Name
protein tyrosine phosphatase type IVA, member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007079.3 Intron NP_009010.2
NM_032611.2 Intron NP_116000.1
XM_005250764.2 Intron XP_005250821.1
XM_006716500.2 Intron XP_006716563.1
XM_011516803.1 Intron XP_011515105.1
XM_011516805.1 Intron XP_011515107.1
XM_017012998.1 Intron XP_016868487.1
XM_017012999.1 Intron XP_016868488.1

View Full Product Details