Product Details

SNP ID
rs62531707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:70580118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAATATCTCCTTCAACAACTCTT[C/T]CATTGCTTCTTGGCATTTTGGCTAA
Phenotype
MIM: 605190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAM1 PubMed Links

Gene Details

Gene
TRAM1
Gene Name
translocation associated membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317804.1 Intron NP_001304733.1
NM_001317805.1 Intron NP_001304734.1
NM_014294.5 Intron NP_055109.1
XM_017013266.1 Intron XP_016868755.1
XM_017013267.1 Intron XP_016868756.1

View Full Product Details