Product Details

SNP ID
rs62515229
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55746710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATTCCTACTAAGCTTTGGAAAA[C/T]AGTAAAATACAGTGGACTAAAGGAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TMEM68 PubMed Links

Gene Details

Gene
TMEM68
Gene Name
transmembrane protein 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286657.1 Intron NP_001273586.1
NM_001286660.1 Intron NP_001273589.1
NM_001286661.1 Intron NP_001273590.1
NM_152417.2 Intron NP_689630.1
XM_005251150.1 Intron XP_005251207.1
XM_011517454.1 Intron XP_011515756.1
XM_011517455.2 Intron XP_011515757.1
XM_017013040.1 Intron XP_016868529.1

View Full Product Details