Product Details

SNP ID
rs61754981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55773649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGAGAAGTGGAGCCGCGTGGCG[C/G]AAATGTTTCTCTTCATTGAGGAGCG
Phenotype
MIM: 606461
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TGS1 PubMed Links

Gene Details

Gene
TGS1
Gene Name
trimethylguanosine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317902.1 204 UTR 5 NP_001304831.1
NM_024831.7 204 Missense Mutation CAA,GAA Q11E NP_079107.6
XM_005251328.2 204 UTR 5 XP_005251385.1
XM_006716485.2 204 Missense Mutation CAA,GAA Q11E XP_006716548.1
XM_017014004.1 204 Missense Mutation CAA,GAA Q11E XP_016869493.1
Gene
TMEM68
Gene Name
transmembrane protein 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286657.1 204 Intron NP_001273586.1
NM_001286660.1 204 Intron NP_001273589.1
NM_001286661.1 204 Intron NP_001273590.1
NM_152417.2 204 Intron NP_689630.1
XM_005251150.1 204 Intron XP_005251207.1
XM_011517454.1 204 Intron XP_011515756.1
XM_011517455.2 204 Intron XP_011515757.1
XM_017013040.1 204 Intron XP_016868529.1

View Full Product Details