Product Details

SNP ID
hCV88978079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56214609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTACCCTCAGTAAGAAGACTG[C/T]TAGAATGCCCTCGGTAACACAGAGG
Phenotype
MIM: 611238 MIM: 603026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD7 PubMed Links

Gene Details

Gene
CHCHD7
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 7
There are no transcripts associated with this gene.

Gene
PLAG1
Gene Name
PLAG1 zinc finger
There are no transcripts associated with this gene.

View Full Product Details