Product Details

SNP ID
rs55782058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32630962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGGCCAAGAATCTGGAACTG[C/T]CATCATTTTCTGGGTGACAATGTTG
Phenotype
MIM: 607798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF1L PubMed Links

Gene Details

Gene
TAF1L
Gene Name
TATA-box binding protein associated factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153809.2 4708 Missense Mutation ACA,GCA T1540A NP_722516.1

View Full Product Details