Product Details

SNP ID
rs59270301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5769160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCGGGTAGACAACTTTGTAATA[A/G]CCCTGAAGAGACTCCACAAAGATTT
Phenotype
MIM: 611156 MIM: 610354
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMP1 PubMed Links

Gene Details

Gene
ERMP1
Gene Name
endoplasmic reticulum metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024896.2 4076 Intron NP_079172.2
XM_005251587.1 4076 Intron XP_005251644.1
XM_017015139.1 4076 Intron XP_016870628.1
Gene
RIC1
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135920.2 4076 Missense Mutation ACC,GCC T1110A NP_001129392.2
NM_001206557.1 4076 Missense Mutation ACC,GCC T1073A NP_001193486.1
NM_020829.3 4076 Missense Mutation ACC,GCC T1110A NP_065880.2
XM_005251523.3 4076 Missense Mutation ACC,GCC T1109A XP_005251580.1
XM_011517967.2 4076 Missense Mutation ACC,GCC T799A XP_011516269.1
XM_011517968.2 4076 Missense Mutation ACC,GCC T573A XP_011516270.1
XM_017014934.1 4076 Missense Mutation ACC,GCC T995A XP_016870423.1
XM_017014935.1 4076 Missense Mutation ACC,GCC T995A XP_016870424.1
XM_017014936.1 4076 Missense Mutation ACC,GCC T995A XP_016870425.1

View Full Product Details