Product Details

SNP ID
rs57315903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114336431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGGTACTGAAGGGGACCATGGT[A/G]GATGGCATCCTGGGCACTTTGTAGC
Phenotype
MIM: 605729 MIM: 138600 MIM: 138610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKNA PubMed Links

Gene Details

Gene
AKNA
Gene Name
AT-hook transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317950.1 4748 UTR 3 NP_001304879.1
NM_001317952.1 4748 UTR 3 NP_001304881.1
NM_030767.5 4748 UTR 3 NP_110394.3
XM_005252244.2 4748 UTR 3 XP_005252301.1
XM_005252245.1 4748 UTR 3 XP_005252302.1
XM_005252247.4 4748 UTR 3 XP_005252304.1
XM_005252248.2 4748 UTR 3 XP_005252305.1
XM_006717294.1 4748 UTR 3 XP_006717357.1
XM_006717295.2 4748 Intron XP_006717358.1
XM_011519063.2 4748 UTR 3 XP_011517365.1
XM_011519064.2 4748 Intron XP_011517366.1
XM_011519065.2 4748 UTR 3 XP_011517367.2
XM_011519066.2 4748 Intron XP_011517368.1
XM_017015172.1 4748 UTR 3 XP_016870661.1
Gene
ORM1
Gene Name
orosomucoid 1
There are no transcripts associated with this gene.

Gene
ORM2
Gene Name
orosomucoid 2
There are no transcripts associated with this gene.

View Full Product Details