Product Details

SNP ID
rs61100236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122675203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTCTCGGTCTGAAGACCAGA[A/G]GCCACTCTTTGCCCTCTTTCTTATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1L3 PubMed Links

Gene Details

Gene
OR1L3
Gene Name
olfactory receptor family 1 subfamily L member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005234.1 74 Missense Mutation AAG,AGG K25R NP_001005234.1

View Full Product Details