Product Details

SNP ID
rs55913281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124356031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGCCCTGTGCTGCACCCAGAT[A/G]CTGCAGCTGGAAGCAGCACTCAGGG
Phenotype
MIM: 604884 MIM: 604030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100129034 PubMed Links

Gene Details

Gene
LOC100129034
Gene Name
uncharacterized LOC100129034
There are no transcripts associated with this gene.

Gene
NEK6
Gene Name
NIMA related kinase 6
There are no transcripts associated with this gene.

Gene
PSMB7
Gene Name
proteasome subunit beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002799.3 Intron NP_002790.1

View Full Product Details