Product Details

SNP ID
rs61119108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124668733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAATTATTCTCTAATATAAATA[C/T]TAATTCAATGAACACACTTATACAG
Phenotype
MIM: 602778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR181A2HG PubMed Links

Gene Details

Gene
MIR181A2HG
Gene Name
MIR181A2 host gene
There are no transcripts associated with this gene.

Gene
NR6A1
Gene Name
nuclear receptor subfamily 6 group A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278546.1 Intron NP_001265475.1
NM_001489.4 Intron NP_001480.3
NM_033334.3 Intron NP_201591.2
XM_005251917.4 Intron XP_005251974.1
XM_005251918.4 Intron XP_005251975.1

View Full Product Details