Product Details
- SNP ID
-
rs61757224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128203516 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCTCATCCCGCTGGTCAACCGG[A/C]TGCAAGACGCCTTCTCTGCCATCGG
- Phenotype
-
MIM: 611420
MIM: 602377
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CIZ1
PubMed Links
Gene Details
- Gene
- CIZ1
- Gene Name
- CDKN1A interacting zinc finger protein 1
- Gene
- DNM1
- Gene Name
- dynamin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005336.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
NP_001005336.1 |
NM_001288737.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
NP_001275666.1 |
NM_001288738.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
NP_001275667.1 |
NM_001288739.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
NP_001275668.1 |
NM_004408.3 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
NP_004399.2 |
XM_005251763.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_005251820.1 |
XM_005251768.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_005251825.1 |
XM_005251769.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_005251826.1 |
XM_006716992.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_006717055.1 |
XM_006716993.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_006717056.1 |
XM_011518335.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_011516637.1 |
XM_011518336.2 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_011516638.1 |
XM_017014369.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_016869858.1 |
XM_017014370.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_016869859.1 |
XM_017014371.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_016869860.1 |
XM_017014372.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_016869861.1 |
XM_017014373.1 |
162 |
Missense Mutation |
ATG,CTG |
M16L |
XP_016869862.1 |
View Full Product Details