Product Details

SNP ID
rs58275320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70771455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGGAGGACTTGAGCTTCTCCA[A/G]TGTACTCTAGTGGGAGGAGAGACAG
Phenotype
MIM: 607506 MIM: 612640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS14 PubMed Links

Gene Details

Gene
ADAMTS14
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
There are no transcripts associated with this gene.

Gene
TBATA
Gene Name
thymus, brain and testes associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318241.1 1616 Missense Mutation ACT,ATT T327I NP_001305170.1
NM_001318242.1 1616 Missense Mutation ACT,ATT T326I NP_001305171.1
NM_001318243.1 1616 Missense Mutation ACT,ATT T325I NP_001305172.1
NM_152710.3 1616 Missense Mutation ACT,ATT T326I NP_689923.3
XM_017015845.1 1616 UTR 3 XP_016871334.1
XM_017015846.1 1616 UTR 3 XP_016871335.1
XM_017015847.1 1616 UTR 3 XP_016871336.1
XM_017015848.1 1616 Silent Mutation CAC,CAT H374H XP_016871337.1
XM_017015849.1 1616 Silent Mutation CTG,TTG L390L XP_016871338.1
XM_017015850.1 1616 Intron XP_016871339.1
XM_017015851.1 1616 Intron XP_016871340.1
XM_017015852.1 1616 Silent Mutation CAC,CAT H340H XP_016871341.1
XM_017015853.1 1616 Missense Mutation ACT,ATT T358I XP_016871342.1
XM_017015854.1 1616 Intron XP_016871343.1
XM_017015855.1 1616 Intron XP_016871344.1
XM_017015856.1 1616 UTR 3 XP_016871345.1
XM_017015857.1 1616 Intron XP_016871346.1
XM_017015858.1 1616 UTR 3 XP_016871347.1
XM_017015859.1 1616 Intron XP_016871348.1
XM_017015860.1 1616 Silent Mutation CAC,CAT H282H XP_016871349.1
XM_017015861.1 1616 Intron XP_016871350.1
XM_017015862.1 1616 Intron XP_016871351.1

View Full Product Details