Product Details

SNP ID
rs55714260
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:52767272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTAACAATAGAATGGATGAAC[G/T]GAGGAATATTAATATAAGGGAACCC
Phenotype
MIM: 154545
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MBL2 PubMed Links

Gene Details

Gene
MBL2
Gene Name
mannose binding lectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000242.2 1745 UTR 3 NP_000233.1
XM_006717861.3 1745 UTR 3 XP_006717924.1
XM_011539816.2 1745 UTR 3 XP_011538118.1

View Full Product Details