Product Details

SNP ID
rs58152081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56359679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTCCCTGTACTCAAGACCCCT[A/G]GGTGTACCTTGGCCTGGAGCTGCTC
Phenotype
MIM: 609177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 Intron NP_001005413.1
NM_007057.3 Intron NP_008988.2
NM_032997.2 Intron NP_127490.1
XM_017015605.1 Intron XP_016871094.1

View Full Product Details