Product Details

SNP ID
rs61757220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21759326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTTCTCAGGCTCTCCTCGTC[A/G]CTGGACTCGTTTTGTTCTGCTATGT
Phenotype
MIM: 611207
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC1 PubMed Links

Gene Details

Gene
DNAJC1
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022365.3 1727 Silent Mutation AGC,AGT S480S NP_071760.2
XM_011519614.2 1727 Intron XP_011517916.1
XM_017016536.1 1727 Intron XP_016872025.1

View Full Product Details