Product Details

SNP ID
rs61918559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93665994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGATTATAGTTTTGAAAACAT[G/T]TATTCTATGCTAACATCTTTTTAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP295 PubMed Links

Gene Details

Gene
CEP295
Gene Name
centrosomal protein 295
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033395.1 Intron NP_203753.1
XM_005274366.1 Intron XP_005274423.1
XM_005274367.2 Intron XP_005274424.1
XM_005274368.2 Intron XP_005274425.1
XM_011543047.1 Intron XP_011541349.1
XM_011543048.1 Intron XP_011541350.1
XM_011543049.1 Intron XP_011541351.1
XM_011543050.2 Intron XP_011541352.1
XM_011543053.2 Intron XP_011541355.1
XM_011543054.2 Intron XP_011541356.1
XM_017018470.1 Intron XP_016873959.1
XM_017018471.1 Intron XP_016873960.1
XM_017018472.1 Intron XP_016873961.1
XM_017018473.1 Intron XP_016873962.1
XM_017018474.1 Intron XP_016873963.1
XM_017018475.1 Intron XP_016873964.1
XM_017018476.1 Intron XP_016873965.1

View Full Product Details