Product Details
- SNP ID
-
rs59859401
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:15972021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTATGTCATACCACATCACGC[A/T]CTGATGGCACGTCAACAGGCAAACA
- Phenotype
-
MIM: 607257
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6073
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59410528] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6073
- Gene Name
- microRNA 6073
There are no transcripts associated with this gene.
- Gene
- SOX6
- Gene Name
- SRY-box 6
View Full Product Details