Product Details

SNP ID
rs57913366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43261555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTATAGGGTTAACACAATAAAC[A/G]ATTCACAGTTGTTAGTTTTAAAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPKP3 PubMed Links

Gene Details

Gene
HNRNPKP3
Gene Name
heterogeneous nuclear ribonucleoprotein K pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details