Product Details

SNP ID
rs60709620
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6977134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGTTTAGCTGTATTAACTTAC[A/C]AGGGAAATGGATTATTCCATCTTCT
Phenotype
MIM: 611531 MIM: 611950 MIM: 610704 MIM: 615642
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
EMG1 PubMed Links

Gene Details

Gene
EMG1
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320049.1 1590 Intron NP_001306978.1
NM_006331.7 1590 Intron NP_006322.4
Gene
LPCAT3
Gene Name
lysophosphatidylcholine acyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005768.5 1590 UTR 3 NP_005759.4
Gene
PHB2
Gene Name
prohibitin 2
There are no transcripts associated with this gene.

Gene
SCARNA12
Gene Name
small Cajal body-specific RNA 12
There are no transcripts associated with this gene.

View Full Product Details