Product Details

SNP ID
rs61748115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120303579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTAGAATGAAGATGAGCTTTGC[A/G]TTGACTTTCAGGTCAGCAAAAGGCC
Phenotype
MIM: 604482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIRT4 PubMed Links

Gene Details

Gene
SIRT4
Gene Name
sirtuin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012240.2 259 Silent Mutation GCA,GCG A6A NP_036372.1
XM_005253865.4 259 Silent Mutation GCA,GCG A6A XP_005253922.1
XM_006719308.2 259 Silent Mutation GCA,GCG A6A XP_006719371.1
XM_006719309.3 259 Silent Mutation GCA,GCG A6A XP_006719372.1

View Full Product Details