Product Details

SNP ID
rs61938670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:117145651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACAGTCCTCACTCGGGACCGAA[C/T]ACGTTCCACAGAGAAATCAGAAGGA
Phenotype
MIM: 609095
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO21 PubMed Links

Gene Details

Gene
FBXO21
Gene Name
F-box protein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015002.2 2187 UTR 3 NP_055817.1
NM_033624.2 2187 UTR 3 NP_296373.1
XM_017019037.1 2187 UTR 3 XP_016874526.1
XM_017019038.1 2187 UTR 3 XP_016874527.1
XM_017019039.1 2187 UTR 3 XP_016874528.1
Gene
TESC-AS1
Gene Name
TESC antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details