Product Details

SNP ID
rs61966753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96092202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGTGCCAGGATTTGGACTCAC[C/T]TGTCCTCAGAAAGATTTCCATGCTC
Phenotype
MIM: 609401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HS6ST3 PubMed Links

Gene Details

Gene
HS6ST3
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153456.3 Intron NP_703157.2
XM_011521076.2 Intron XP_011519378.1
XM_017020543.1 Intron XP_016876032.1

View Full Product Details