Product Details

SNP ID
rs57239631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40945995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAACAATCTGTGAATTGCATGGC[A/G]AGGATGGAGAGTGAGATAGCTGGAG
Phenotype
MIM: 614587 MIM: 605185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHAC1 PubMed Links

Gene Details

Gene
CHAC1
Gene Name
ChaC glutathione specific gamma-glutamylcyclotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142776.1 Intron NP_001136248.1
NM_024111.3 Intron NP_077016.2
XM_017022563.1 Intron XP_016878052.1
XM_017022564.1 Intron XP_016878053.1
XM_017022565.1 Intron XP_016878054.1
Gene
DLL4
Gene Name
delta like canonical Notch ligand 4
There are no transcripts associated with this gene.

View Full Product Details