Product Details

SNP ID
rs56323146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45094608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTTGAAGAAGGGCTCGAAGGG[C/G]GGACGGCCAAAGTGGGTGATGGAGC
Phenotype
MIM: 606759
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014080.4 4682 Silent Mutation CCC,CCG P1493P NP_054799.4
XM_005254421.2 4682 Silent Mutation CCC,CCG P1493P XP_005254478.1

View Full Product Details