Product Details

SNP ID
rs56176906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52192869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGAAAGGTATTAAAAAATAAAA[A/T]AATTATGGTGAATATGTTGGCAGGT
Phenotype
MIM: 604447 MIM: 610022
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GNB5 PubMed Links

Gene Details

Gene
GNB5
Gene Name
G protein subunit beta 5
There are no transcripts associated with this gene.

Gene
LOC100129973
Gene Name
uncharacterized LOC100129973
There are no transcripts associated with this gene.

Gene
MYO5C
Gene Name
myosin VC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018728.3 6424 UTR 3 NP_061198.2
XM_011521781.2 6424 UTR 3 XP_011520083.1
XM_017022408.1 6424 UTR 3 XP_016877897.1
XM_017022409.1 6424 UTR 3 XP_016877898.1
XM_017022410.1 6424 UTR 3 XP_016877899.1

View Full Product Details