Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105192.2 | 4047 | UTR 3 | NP_001098662.1 | ||
NM_001282979.1 | 4047 | UTR 3 | NP_001269908.1 | ||
NM_001282980.1 | 4047 | UTR 3 | NP_001269909.1 | ||
NM_001282981.1 | 4047 | UTR 3 | NP_001269910.1 | ||
NM_001282982.1 | 4047 | UTR 3 | NP_001269911.1 | ||
NM_005078.3 | 4047 | UTR 3 | NP_005069.2 | ||
NM_020908.2 | 4047 | UTR 3 | NP_065959.1 | ||
XM_005254622.4 | 4047 | UTR 3 | XP_005254679.1 | ||
XM_005254623.3 | 4047 | UTR 3 | XP_005254680.1 | ||
XM_005254625.3 | 4047 | UTR 3 | XP_005254682.1 | ||
XM_005254628.3 | 4047 | UTR 3 | XP_005254685.1 | ||
XM_005254633.2 | 4047 | UTR 3 | XP_005254690.1 | ||
XM_006720665.3 | 4047 | UTR 3 | XP_006720728.1 | ||
XM_011521976.2 | 4047 | UTR 3 | XP_011520278.1 | ||
XM_011521977.2 | 4047 | UTR 3 | XP_011520279.1 | ||
XM_011521978.2 | 4047 | UTR 3 | XP_011520280.1 | ||
XM_011521979.2 | 4047 | UTR 3 | XP_011520281.1 | ||
XM_011521980.2 | 4047 | UTR 3 | XP_011520282.1 | ||
XM_011521981.1 | 4047 | Intron | XP_011520283.1 | ||
XM_011521982.2 | 4047 | UTR 3 | XP_011520284.1 | ||
XM_011521983.2 | 4047 | UTR 3 | XP_011520285.1 | ||
XM_017022532.1 | 4047 | Intron | XP_016878021.1 |