Product Details
- SNP ID
-
rs56258098
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78628975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGTTAGCAACTTACACTCTGGT[A/C]TTCATCGTCATTCTTCATGTGCTGG
- Phenotype
-
MIM: 118503
MIM: 118509
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CHRNA3
PubMed Links
Gene Details
- Gene
- CHRNA3
- Gene Name
- cholinergic receptor nicotinic alpha 3 subunit
There are no transcripts associated with this gene.
- Gene
- CHRNB4
- Gene Name
- cholinergic receptor nicotinic beta 4 subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000750.3 |
1190 |
Missense Mutation |
GAC,TAC |
D444Y |
NP_000741.1 |
NM_001256567.1 |
1190 |
Intron |
|
|
NP_001243496.1 |
XM_011521186.2 |
1190 |
Missense Mutation |
GAC,TAC |
D441Y |
XP_011519488.1 |
XM_011521187.2 |
1190 |
Missense Mutation |
GAC,TAC |
D441Y |
XP_011519489.1 |
XM_011521190.2 |
1190 |
Missense Mutation |
GAC,TAC |
D370Y |
XP_011519492.1 |
XM_011521191.2 |
1190 |
Missense Mutation |
GAC,TAC |
D370Y |
XP_011519493.1 |
XM_011521192.2 |
1190 |
Missense Mutation |
GAC,TAC |
D262Y |
XP_011519494.1 |
XM_017021885.1 |
1190 |
Missense Mutation |
GAC,TAC |
D414Y |
XP_016877374.1 |
XM_017021886.1 |
1190 |
Intron |
|
|
XP_016877375.1 |
XM_017021887.1 |
1190 |
Missense Mutation |
GAC,TAC |
D444Y |
XP_016877376.1 |
XM_017021888.1 |
1190 |
Missense Mutation |
GAC,TAC |
D444Y |
XP_016877377.1 |
XM_017021889.1 |
1190 |
Missense Mutation |
GAC,TAC |
D444Y |
XP_016877378.1 |
View Full Product Details