Product Details

SNP ID
rs62019465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84755846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGTAAGATGCTTTTGTTAGATA[C/T]GCATTCAAACAGGTGGATTAGGCTA
Phenotype
MIM: 613624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF592 PubMed Links

Gene Details

Gene
ZNF592
Gene Name
zinc finger protein 592
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014630.2 Intron NP_055445.2
XM_005254996.3 Intron XP_005255053.1
XM_011522246.2 Intron XP_011520548.1
XM_011522247.2 Intron XP_011520549.1
XM_011522248.2 Intron XP_011520550.1
XM_017022734.1 Intron XP_016878223.1

View Full Product Details