Product Details

SNP ID
rs57288618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:16150599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCGGGCACAGTCCATCACGGAG[C/T]GCAGGCGGTGGGCAATGAGCAGCAC
Phenotype
MIM: 158343 MIM: 603234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC1 PubMed Links

Gene Details

Gene
ABCC1
Gene Name
ATP binding cassette subfamily C member 1
There are no transcripts associated with this gene.

Gene
ABCC6
Gene Name
ATP binding cassette subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079528.3 4395 Intron NP_001072996.1
NM_001171.5 4395 Missense Mutation CAC,CGC H1461R NP_001162.4
XM_011522479.2 4395 Missense Mutation CAC,CGC H1450R XP_011520781.1
XM_011522480.1 4395 Missense Mutation CAC,CGC H1347R XP_011520782.1
XM_011522481.2 4395 Missense Mutation CAC,CGC H1347R XP_011520783.1
XM_011522482.2 4395 Intron XP_011520784.1
XM_017023212.1 4395 Missense Mutation CAC,CGC H1405R XP_016878701.1
XM_017023213.1 4395 Missense Mutation CAC,CGC H1347R XP_016878702.1
XM_017023214.1 4395 Intron XP_016878703.1

View Full Product Details