Product Details

SNP ID
rs61553825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:16150667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCGAGCATGGCCTGCATCTG[C/T]AGCTCCGTGCCAGGGTCCACGGCAG
Phenotype
MIM: 158343 MIM: 603234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC1 PubMed Links

Gene Details

Gene
ABCC1
Gene Name
ATP binding cassette subfamily C member 1
There are no transcripts associated with this gene.

Gene
ABCC6
Gene Name
ATP binding cassette subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079528.3 4327 Intron NP_001072996.1
NM_001171.5 4327 Silent Mutation CTA,CTG L1438L NP_001162.4
XM_011522479.2 4327 Silent Mutation CTA,CTG L1427L XP_011520781.1
XM_011522480.1 4327 Silent Mutation CTA,CTG L1324L XP_011520782.1
XM_011522481.2 4327 Silent Mutation CTA,CTG L1324L XP_011520783.1
XM_011522482.2 4327 Intron XP_011520784.1
XM_017023212.1 4327 Silent Mutation CTA,CTG L1382L XP_016878701.1
XM_017023213.1 4327 Silent Mutation CTA,CTG L1324L XP_016878702.1
XM_017023214.1 4327 Intron XP_016878703.1

View Full Product Details