Product Details
- SNP ID
-
rs60493962
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:54285084 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCGCCCCAGCCAGGGACAGCTCA[A/G]GCTCGGTGGCCGCGCCGTCTAAGTT
- Phenotype
-
MIM: 612985
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IRX3
PubMed Links
Gene Details
- Gene
- IRX3
- Gene Name
- iroquois homeobox 3
View Full Product Details