Product Details

SNP ID
rs59083128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75320247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAACCAAAGTGAGAGGGTCACAG[A/G]GTGACCCGGCCAGGTGTTCTGCACT
Phenotype
MIM: 108355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRB2 PubMed Links

Gene Details

Gene
GRB2
Gene Name
growth factor receptor bound protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002086.4 1010 UTR 3 NP_002077.1
NM_203506.2 1010 UTR 3 NP_987102.1

View Full Product Details