Product Details

SNP ID
rs56895494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75517538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGTGAGGGCTCATAAGCTGAG[A/C]TGTTGGCCCCACTTCCAGGGCTCCA
Phenotype
MIM: 612185 MIM: 608755
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASKIN2 PubMed Links

Gene Details

Gene
CASKIN2
Gene Name
CASK interacting protein 2
There are no transcripts associated with this gene.

Gene
LLGL2
Gene Name
LLGL2, scribble cell polarity complex component
There are no transcripts associated with this gene.

Gene
TSEN54
Gene Name
tRNA splicing endonuclease subunit 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207346.2 Intron NP_997229.2
XM_005257229.3 Intron XP_005257286.1

View Full Product Details