Product Details

SNP ID
rs55664828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76386220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATAGCTGCCGGTCTCCCTGCA[A/G]AGCGGCGGAACCACGCGCGGGAGCT
Phenotype
MIM: 603730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPHK1 PubMed Links

Gene Details

Gene
SPHK1
Gene Name
sphingosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142601.1 Intron NP_001136073.1
NM_001142602.1 Intron NP_001136074.1
NM_021972.3 Intron NP_068807.2
NM_182965.2 Intron NP_892010.2
XM_005257766.2 Intron XP_005257823.1
Gene
UBE2O
Gene Name
ubiquitin conjugating enzyme E2 O
There are no transcripts associated with this gene.

View Full Product Details