Product Details

SNP ID
rs62079522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82394358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCTTAAAGCAGGACTCAGCAC[A/G]GCAACCAAGGAATCAACAATAAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OGFOD3 PubMed Links

Gene Details

Gene
OGFOD3
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024648.2 1161 Intron NP_078924.1
NM_175902.4 1161 UTR 3 NP_787098.3

View Full Product Details