Product Details

SNP ID
rs61758747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82658061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCTTCTTAAGGCAATGGGGAG[C/T]GGGAGCTTGTCCACCAGGTGCACCG
Phenotype
MIM: 609226
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 877 Silent Mutation CCA,CCG P213P NP_006813.1
XM_005256334.4 877 Silent Mutation CCA,CCG P153P XP_005256391.1
XM_005256335.3 877 Intron XP_005256392.1
XM_006722271.3 877 Silent Mutation CCA,CCG P173P XP_006722334.1
XM_011523528.2 877 Silent Mutation CCA,CCG P196P XP_011521830.1
XM_017024042.1 877 Intron XP_016879531.1
XM_017024043.1 877 Silent Mutation CCA,CCG P153P XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details