Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321164.1 | 3075 | Intron | NP_001308093.1 | ||
NM_001321166.1 | 3075 | Intron | NP_001308095.1 | ||
NM_001321167.1 | 3075 | Intron | NP_001308096.1 | ||
NM_001321168.1 | 3075 | Intron | NP_001308097.1 | ||
NM_014859.5 | 3075 | Intron | NP_055674.4 | ||
XM_005256891.1 | 3075 | Intron | XP_005256948.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001165962.1 | 3075 | UTR 3 | NP_001159434.1 | ||
NM_018127.6 | 3075 | UTR 3 | NP_060597.4 | ||
NM_173717.1 | 3075 | UTR 3 | NP_776065.1 |