Product Details
- SNP ID
-
rs55880817
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1842817 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCCCTCAGGCCATCATGCAGAA[A/G]GGGGATACAAACATAAAGCCCATCC
- Phenotype
-
MIM: 179835
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPA1
PubMed Links
Gene Details
- Gene
- RPA1
- Gene Name
- replication protein A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002945.3 |
163 |
Silent Mutation |
AAA,AAG |
K16K |
NP_002936.1 |
View Full Product Details