Product Details

SNP ID
rs59995125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17812063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCAGCACATCCATCAGCTGAAG[A/G]CACAAAACCCAGATTATAAATAATT
Phenotype
MIM: 613486 MIM: 607642 MIM: 184756
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR33B PubMed Links

Gene Details

Gene
MIR33B
Gene Name
microRNA 33b
There are no transcripts associated with this gene.

Gene
MIR6777
Gene Name
microRNA 6777
There are no transcripts associated with this gene.

Gene
RAI1
Gene Name
retinoic acid induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030665.3 4197 Intron NP_109590.3
XM_017024025.1 4197 Intron XP_016879514.1
XM_017024026.1 4197 Intron XP_016879515.1
XM_017024027.1 4197 Intron XP_016879516.1
XM_017024028.1 4197 Intron XP_016879517.1
Gene
SREBF1
Gene Name
sterol regulatory element binding transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005291.2 4197 UTR 3 NP_001005291.1
NM_001321096.2 4197 Intron NP_001308025.1
NM_004176.4 4197 UTR 3 NP_004167.3
XM_005256772.4 4197 Intron XP_005256829.1
XM_017024970.1 4197 Intron XP_016880459.1
XM_017024971.1 4197 Intron XP_016880460.1

View Full Product Details