Product Details

SNP ID
rs61754585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:745039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGGGTTTCCAAGGCTAAAACGT[A/G]GAGTGGCTCACAGACCTCCGGGTGG
Phenotype
MIM: 611627 MIM: 606969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015721.2 3341 Missense Mutation CAC,TAC H1002Y NP_056536.2
XM_005256667.4 3341 Missense Mutation CAC,TAC H1006Y XP_005256724.1
XM_005256670.4 3341 Missense Mutation CAC,TAC H991Y XP_005256727.1
XM_011523910.2 3341 Missense Mutation CAC,TAC H1006Y XP_011522212.1
XM_011523911.2 3341 Missense Mutation CAC,TAC H1006Y XP_011522213.1
XM_011523912.2 3341 Missense Mutation CAC,TAC H991Y XP_011522214.1
XM_011523913.2 3341 Missense Mutation CAC,TAC H991Y XP_011522215.1
XM_017024709.1 3341 Missense Mutation CAC,TAC H1006Y XP_016880198.1

View Full Product Details