Product Details
- SNP ID
-
rs62068292
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1006167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCAGGTAGTAGAGGAGGACCTG[C/G]ACCTGTGGGGAGACAGGAAGGCGGA
- Phenotype
-
MIM: 600365
MIM: 607251
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ABR
PubMed Links
Gene Details
- Gene
- ABR
- Gene Name
- active BCR-related
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001092.4 |
2695 |
Silent Mutation |
GTC,GTG |
V794V |
NP_001083.2 |
NM_001159746.2 |
2695 |
Silent Mutation |
GTC,GTG |
V785V |
NP_001153218.1 |
NM_001256847.2 |
2695 |
Silent Mutation |
GTC,GTG |
V282V |
NP_001243776.1 |
NM_001282149.1 |
2695 |
Silent Mutation |
GTC,GTG |
V613V |
NP_001269078.1 |
NM_001322840.1 |
2695 |
Silent Mutation |
GTC,GTG |
V785V |
NP_001309769.1 |
NM_001322841.1 |
2695 |
Silent Mutation |
GTC,GTG |
V1090V |
NP_001309770.1 |
NM_001322842.1 |
2695 |
Intron |
|
|
NP_001309771.1 |
NM_021962.4 |
2695 |
Silent Mutation |
GTC,GTG |
V831V |
NP_068781.2 |
XM_011523810.2 |
2695 |
Silent Mutation |
GTC,GTG |
V850V |
XP_011522112.1 |
XM_011523811.2 |
2695 |
Intron |
|
|
XP_011522113.2 |
XM_011523812.2 |
2695 |
Silent Mutation |
GTC,GTG |
V837V |
XP_011522114.1 |
XM_011523813.2 |
2695 |
Silent Mutation |
GTC,GTG |
V813V |
XP_011522115.1 |
XM_011523814.2 |
2695 |
Silent Mutation |
GTC,GTG |
V804V |
XP_011522116.1 |
XM_011523815.2 |
2695 |
Silent Mutation |
GTC,GTG |
V804V |
XP_011522117.1 |
XM_017024539.1 |
2695 |
Silent Mutation |
GTC,GTG |
V1109V |
XP_016880028.1 |
XM_017024540.1 |
2695 |
Silent Mutation |
GTC,GTG |
V1076V |
XP_016880029.1 |
XM_017024541.1 |
2695 |
Intron |
|
|
XP_016880030.1 |
XM_017024542.1 |
2695 |
Silent Mutation |
GTC,GTG |
V635V |
XP_016880031.1 |
XM_017024543.1 |
2695 |
Silent Mutation |
GTC,GTG |
V632V |
XP_016880032.1 |
- Gene
- TIMM22
- Gene Name
- translocase of inner mitochondrial membrane 22 homolog (yeast)
There are no transcripts associated with this gene.
View Full Product Details