Product Details

SNP ID
rs61749667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8003336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTGGCAGGCGGTCCCCGCTTC[C/G]AGGTAGCGCTGCTGCCCGAGCCTTG
Phenotype
MIM: 600179
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GUCY2D PubMed Links

Gene Details

Gene
GUCY2D
Gene Name
guanylate cyclase 2D, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000180.3 289 Missense Mutation CAG,GAG Q97E NP_000171.1
XM_011523816.1 289 Missense Mutation CAG,GAG Q97E XP_011522118.1

View Full Product Details